Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172112.1 | NP_001165583.1 | 6 | 745 | ||
NM_145428.2 | NP_663403.1 | 6 | 716 | |||
GenBank mRNA | AK052209.1 | - | 6 | 668 | ||
AK139084.1 | - | 6 | 697 | |||
AK144057.1 | - | 6 | 651 | |||
BC003479.1 | - | 6 | 671 | 2655855 | ||
JX196872.1 | - | 6 | 635 |