Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300ld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145437.2 | NP_663412.1 | 4 | 912 | ||
GenBank mRNA | AK028662.1 | - | 4 | 924 | ||
AK041292.1 | - | 4 | 948 | |||
BC019814.1 | - | 4 | 912 | 5125383 |