Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9030617O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145448.4 | NP_663423.2 | 8 | 1336 | ||
GenBank mRNA | AK033551.1 | - | 8 | 1365 | ||
AK038436.1 | - | 8 | 1338 | |||
AK081256.1 | - | 1373 | ||||
AK152622.1 | - | 8 | 1365 | |||
BC021385.1 | - | 9 | 1321 | 5064508 | ||
BC066161.1 | - | 9 | 1410 | 30093441 |