Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145452.3 | NP_663427.2 | 8 | 1773 | ||
GenBank mRNA | AK042801.1 | - | 1 | 1132 | ||
AK133354.1 | - | 4 | 333 | |||
BC013637.1 | - | 6 | 519 | 3498774 | ||
BC065082.1 | - | 8 | 1316 |