Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145460.2 | NP_663435.2 | 8 | 1821 | ||
GenBank mRNA | AK031978.1 | - | 7 | 1450 | ||
AK032160.1 | - | 8 | 1624 | |||
AK075924.1 | - | 7 | 1447 | |||
AK139884.1 | - | 8 | 1821 | |||
BC019806.1 | - | 8 | 1597 | 4018387 |