Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145465.2 | NP_663440.1 | 11 | 1935 | ||
GenBank mRNA | AK154596.1 | - | 11 | 2040 | ||
AK154959.1 | - | 11 | 1936 | |||
AK164377.1 | - | 1 | 133 | |||
BC004650.1 | - | 11 | 1767 | 3482980 | ||
BY730101.1 | - | 1 | 133 |