Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 2 | 218 | ||
GenBank mRNA | AK039627.1 | - | 2 | 231 | ||
AK146872.1 | - | 2 | 219 | |||
AK154226.1 | - | 2 | 240 | |||
AK154602.1 | - | 2 | 260 | |||
AK155459.1 | - | 2 | 232 | |||
AK155770.1 | - | 2 | 261 | |||
BC066009.1 | - | 2 | 218 | 5708486 |