Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 13 | 3064 | ||
GenBank mRNA | AK146872.1 | - | 13 | 3065 | ||
AK154226.1 | - | 13 | 3087 | |||
AK154602.1 | - | 13 | 3109 | |||
AK155459.1 | - | 13 | 3079 | |||
AK155770.1 | - | 13 | 3107 | |||
AK207869.1 | - | 1 | 282 | |||
BC023106.1 | - | 10 | 2460 | 4016550 | ||
BC066009.1 | - | 13 | 3064 | 5708486 |