Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145515.2 | NP_663490.2 | 3 | 370 | ||
GenBank mRNA | AB185282.1 | - | 3 | 293 | ||
AF453686.1 | - | 3 | 300 | |||
AK173181.1 | - | 2 | 220 | |||
BC112400.1 | - | 3 | 299 | 40059725 | ||
CJ179387.1 | - | 3 | 391 |