Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005870.4 | NP_005861.2 | 4 | 416 | ||
XM_011534868.2 | XP_011533170.1 | 3 | 296 | |||
GenBank mRNA | AF153608.1 | - | 4 | 354 | ||
AK311748.1 | - | 4 | 337 | |||
AY550970.1 | - | 4 | 336 | |||
CR457035.1 | - | 4 | 320 | |||
DB171768.1 | - | 4 | 418 | |||
U78303.1 | - | 4 | 351 | |||
U96915.1 | - | 4 | 336 |