Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145523.3 | NP_663498.1 | 8 | 918 | ||
GenBank mRNA | AF518325.1 | - | 8 | 902 | ||
AK033740.1 | - | 8 | 951 | |||
AK046347.1 | - | 8 | 960 | |||
AK049317.1 | - | 8 | 919 | |||
AK050540.1 | - | 8 | 960 | |||
AK078915.1 | - | 8 | 919 | |||
BC021450.1 | - | 8 | 912 | 5044040 |