Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145539.2 | NP_663514.2 | 3, 4 | 609 | ||
GenBank mRNA | AK148484.1 | - | 3, 4 | 577 | ||
AK165731.1 | - | 3, 4 | 577 | |||
BC010814.1 | - | 3, 4 | 560 | 4211816 | ||
Z47778.1 | - | 1, 2 | 29 |