Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040690.2 | NP_001035780.1 | 7 | 884 | ||
NM_001286759.1 | NP_001273688.1 | 7 | 884 | |||
NM_145544.3 | NP_663519.2 | 6 | 737 | |||
GenBank mRNA | AK052125.1 | - | 6 | 685 | ||
AK154852.1 | - | 7 | 870 | |||
AK155793.1 | - | 7 | 870 | |||
AK160989.1 | - | 7 | 820 | |||
AK164159.1 | - | 7 | 796 | |||
AK166315.1 | - | 7 | 817 | |||
AK172052.1 | - | 7 | 886 | |||
BC011279.1 | - | 6 | 606 | 4008751 |