Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205156.1 | NP_001192085.1 | 7 | 821 | ||
NM_145550.3 | NP_663525.1 | 8 | 902 | |||
GenBank mRNA | AK048749.1 | - | 2 | 1673 | ||
AK081205.1 | - | 8 | 889 | |||
AK157536.1 | - | 8 | 889 | |||
AK214559.1 | - | 3 | 371 | |||
BC009080.1 | - | 8 | 834 | 2655774 |