Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145564.3 | NP_663539.1 | 12 | 3030 | ||
GenBank mRNA | AB093270.1 | - | 12 | 3001 | ||
AK013157.1 | - | 1 | 522 | |||
AK048131.1 | - | 12 | 2988 | |||
AK048542.1 | - | 12 | 2988 | |||
AK049723.1 | - | 12 | 3031 | |||
BC026383.1 | - | 3 | 1649 |