Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145569.4 | NP_663544.1 | 9 | 2097 | ||
GenBank mRNA | AK034757.1 | - | 9 | 2099 | ||
AK052764.1 | - | 9 | 2098 | |||
AK144189.1 | - | 9 | 2101 | |||
AK145407.1 | - | 9 | 2096 | |||
AK151281.1 | - | 8 | 2682 | |||
AK160869.1 | - | 8 | 2680 | |||
AK164137.1 | - | 9 | 2097 | |||
AK166671.1 | - | 9 | 2097 | |||
AK168155.1 | - | 9 | 2094 | |||
AK169705.1 | - | 9 | 2093 | |||
AK199984.1 | - | 1 | 71 | |||
AK205713.1 | - | 1 | 71 | |||
AK209635.1 | - | 1 | 70 | |||
AK215987.1 | - | 1 | 71 | |||
BC058360.1 | - | 8 | 2658 | 6821781 |