Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145571.2 | NP_663546.1 | 3, 4 | 498 | ||
GenBank mRNA | AK028416.1 | - | 3, 4 | 521 | ||
AK158228.1 | - | 3, 4 | 481 | |||
AK167515.1 | - | 3, 4 | 493 | |||
AK170691.1 | - | 3, 4 | 492 | |||
AK172205.1 | - | 3, 4 | 501 | |||
AK216221.1 | - | 2, 3 | 189 | |||
BC009149.1 | - | 3, 4 | 498 | 2651599 | ||
BC033463.1 | - | 3, 4 | 452 | 3500664 |