Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145573.2 | NP_663548.2 | 8 | 881 | ||
GenBank mRNA | AK077677.1 | - | 8 | 882 | ||
AK170480.1 | - | 8 | 881 | |||
AK212599.1 | - | 1 | 51 | |||
BC019964.1 | - | 8 | 859 | 4158657 |