Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145827.3 | NP_665826.1 | 4 | 932 | ||
GenBank mRNA | AF486632.1 | - | 5 | 828 | ||
AY337285.1 | - | 3 | 718 | |||
AY355340.1 | - | 3 | 706 | |||
AY495376.1 | - | 4 | 932 | |||
AY495377.1 | - | 5 | 828 | |||
BC116174.1 | - | 4 | 892 | 40048724 | ||
BC116175.1 | - | 4 | 892 | 40048725 | ||
KF032621.1 | - | 3 | 708 |