Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145828.3 | NP_665827.2 | 5 | 1138 | ||
GenBank mRNA | AJ291751.2 | - | 5 | 1055 | ||
AK083482.1 | - | 5 | 1107 | |||
AK086804.1 | - | 5 | 1070 | |||
AK150521.1 | - | 5 | 1091 | |||
AK153332.1 | - | 5 | 1091 | |||
AK158056.1 | - | 5 | 1138 | |||
AK177270.1 | - | 2 | 156 | |||
BC034082.1 | - | 4 | 657 | 4239646 |