Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145831.3 | NP_665830.1 | 3, 4 | 1020 | ||
GenBank mRNA | AF539811.1 | - | 3, 4 | 1020 | ||
AK044891.1 | - | 3, 4 | 1021 | |||
AK140337.1 | - | 3, 4 | 1012 | |||
AK140347.1 | - | 3, 4 | 1015 |