Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145963.2 | NP_666075.1 | 3 | 1925 | ||
GenBank mRNA | AK030711.1 | - | 3 | 1935 | ||
AK044725.1 | - | 3 | 1906 | |||
AK080732.1 | - | 3 | 1914 | |||
BC022700.1 | - | 1 | 1024 | |||
BC029692.1 | - | 3 | 1865 | 5363132 | ||
BC031753.1 | - | 3 | 1925 | 4503027 |