Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290539.1 | NP_001277468.1 | 3 | 571 | ||
NM_145967.2 | NP_666079.2 | 3 | 571 | |||
GenBank mRNA | AK032473.1 | - | 3 | 570 | ||
AK033048.1 | - | 3 | 573 | |||
AK079096.1 | - | 493 | ||||
AK137363.1 | - | 3 | 493 | |||
BC027127.1 | - | 3 | 523 | 5361611 | ||
BM963943.1 | - | 3 | 231 |