Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134465.2 | NP_001127937.1 | 13 | 1257 | ||
NM_001285466.1 | NP_001272395.1 | 11 | 1103 | |||
NM_001285467.1 | NP_001272396.1 | 13 | 1318 | |||
NM_145969.4 | NP_666081.1 | 13 | 1257 | |||
GenBank mRNA | AK033036.1 | - | 11 | 1105 | ||
AK054197.1 | - | 13 | 1201 | |||
AK054295.1 | - | 13 | 1258 | |||
AK080327.1 | - | 3 | 198 | |||
AK131163.1 | - | 13 | 1229 | |||
AK132219.1 | - | 13 | 1254 | |||
AK135953.1 | - | 13 | 1269 | |||
AK161908.1 | - | 13 | 1227 | |||
AK163396.1 | - | 13 | 1317 | |||
BC025603.1 | - | 13 | 1252 | 5250476 |