Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244755.1 | NP_001231684.1 | 2 | 118 | ||
NM_005752.4 | NP_005743.4 | 3 | 339 | |||
GenBank mRNA | AA399390.1 | - | 2 | 119 | ||
AK313316.1 | - | 3 | 339 | |||
AY358376.1 | - | 3 | 291 | |||
BC104414.2 | - | 3 | 328 | 40003144 | ||
BC104415.2 | - | 3 | 328 | 40003149 | ||
DA998903.1 | - | 3 | 339 |