Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005754.2 | NP_005745.1 | 7, 8 | 908 | ||
NM_198395.1 | NP_938405.1 | 7, 8 | 877 | |||
XM_006714749.3 | XP_006714812.1 | 7, 8 | 897 | |||
XM_006714750.3 | XP_006714813.1 | 7, 8 | 836 | |||
XM_017008922.1 | XP_016864411.1 | 7, 8 | 1183 | |||
XM_017008923.1 | XP_016864412.1 | 7, 8 | 1152 | |||
GenBank mRNA | AK222606.1 | - | 7, 8 | 869 | ||
AK300098.1 | - | 6, 7 | 778 | |||
AK303580.1 | - | 7, 8 | 965 | |||
AK312572.1 | - | 7, 8 | 871 | |||
BC006997.1 | - | 7, 8 | 871 | 3950738 | ||
BC108278.1 | - | 7, 8 | 816 | 6051310 | ||
BT019384.1 | - | 6, 7 | 737 | |||
BX647869.1 | - | 6, 7 | 779 | |||
BX648012.1 | - | 7, 8 | 883 | |||
CR536535.1 | - | 6, 7 | 737 | |||
U32519.1 | - | 7, 8 | 858 |