Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 13 | 2936 | ||
NM_001271357.1 | NP_001258286.1 | 13 | 2679 | |||
NM_146018.2 | NP_666130.1 | 13 | 2976 | |||
GenBank mRNA | AK039106.1 | - | 13 | 2764 | ||
AK080933.1 | - | 13 | 2761 | |||
AK136071.1 | - | 13 | 2667 | |||
AK161460.1 | - | 13 | 3019 | |||
BC025820.1 | - | 13 | 2710 | 5099493 | ||
BC027276.1 | - | 1 | 749 |