Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146026.1 | NP_666138.1 | 3 | 403 | ||
GenBank mRNA | AK077279.1 | - | 2 | 376 | ||
AK087566.1 | - | 255 | ||||
AK090284.1 | - | 2 | 405 | |||
BC022765.1 | - | 3 | 403 | 4925830 | ||
BC024510.1 | - | 2 | 287 | 4983784 | ||
BC034192.1 | - | 2 | 333 | 4984606 | ||
BC034213.1 | - | 2 | 382 | 4989688 | ||
BC036171.1 | - | 2 | 350 | 4988054 |