Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146036.1 | NP_666148.1 | 4 | 571 | ||
GenBank mRNA | AK076069.1 | - | 4 | 581 | ||
AK165289.1 | - | 4 | 589 | |||
AK166644.1 | - | 4 | 544 | |||
AK210244.1 | - | 2 | 137 | |||
BC023857.1 | - | 4 | 538 | 5346745 | ||
BC025552.1 | - | 4 | 571 | 5320590 | ||
BC036967.1 | - | 1 | 1139 |