Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146045.1 | NP_666157.1 | 4 | 685 | ||
GenBank mRNA | AK141739.1 | - | 4 | 649 | ||
AK146855.1 | - | 4 | 732 | |||
AK171600.1 | - | 4 | 701 | |||
BC027195.1 | - | 4 | 685 | 4224150 | ||
BC055703.1 | - | 4 | 900 | 6403624 |