Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 7 | 1075 | ||
GenBank mRNA | AK032295.1 | - | 7 | 1038 | ||
AK048357.1 | - | 7 | 972 | |||
AK156203.1 | - | 7 | 966 | |||
AK165939.1 | - | 7 | 936 | |||
AK177383.1 | - | 2 | 64 | |||
BC026562.1 | - | 7 | 776 | 5028264 |