Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 8 | 1197 | ||
GenBank mRNA | AK032295.1 | - | 8 | 1160 | ||
AK048357.1 | - | 8 | 1094 | |||
AK156203.1 | - | 8 | 1088 | |||
AK165939.1 | - | 8 | 1058 | |||
AK177383.1 | - | 3 | 188 | |||
BC026562.1 | - | 8 | 898 | 5028264 |