Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 10, 11 | 1382 | ||
GenBank mRNA | AK032295.1 | - | 10, 11 | 1346 | ||
AK048357.1 | - | 10, 11 | 1279 | |||
AK156203.1 | - | 10, 11 | 1273 | |||
AK165939.1 | - | 10, 11 | 1243 | |||
AK177383.1 | - | 5, 6 | 373 | |||
AK181076.1 | - | 2, 3 | 104 | |||
BC026562.1 | - | 10, 11 | 1083 | 5028264 |