Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146057.3 | NP_666169.1 | 2, 3 | 304 | ||
GenBank mRNA | AK014991.1 | - | 2, 3 | 196 | ||
AK152216.1 | - | 2, 3 | 271 | |||
AK152884.1 | - | 2, 3 | 281 | |||
AK167579.1 | - | 2, 3 | 228 | |||
AK167688.1 | - | 2, 3 | 168 | |||
AK169162.1 | - | 2, 3 | 236 | |||
AK171259.1 | - | 2, 3 | 298 | |||
AK197429.1 | - | 1, 2 | 62 | |||
BC010828.1 | - | 2, 3 | 240 | 4220180 | ||
BC024876.1 | - | 2, 3 | 268 | 3155209 | ||
BC057669.1 | - | 2, 3 | 230 | 2655176 | ||
BY272917.1 | - | 2, 3 | 305 |