Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145874.1 | NP_001139346.1 | 2 | 751 | ||
NM_146071.2 | NP_666183.2 | 2 | 751 | |||
GenBank mRNA | AK085640.1 | - | 2 | 718 | ||
AK142548.1 | - | 2 | 752 | |||
AK165477.1 | - | 2 | 743 | |||
BC026367.1 | - | 2 | 752 | 4037422 |