Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005778.3 | NP_005769.1 | 8 | 770 | ||
NR_036627.2 | 7 | 696 | ||||
XM_006712917.1 | XP_006712980.1 | 8 | 770 | |||
XM_011533262.1 | XP_011531564.1 | 8 | 770 | |||
XM_017005503.1 | XP_016860992.1 | 8 | 752 | |||
GenBank mRNA | AB208813.1 | - | 7 | 3186 | ||
AF091263.1 | - | 8 | 743 | |||
AF103802.4 | - | 8 | 721 | |||
AK297249.1 | - | 7 | 650 | |||
AK297445.1 | - | 8 | 625 | |||
AK302766.1 | - | 7 | 578 | |||
AK314032.1 | - | 8 | 725 | |||
AU129811.1 | - | 1 | 37 | |||
BC002957.1 | - | 8 | 716 | |||
KU178505.1 | - | 7 | 595 | |||
KU178506.1 | - | 6 | 521 | |||
U23946.1 | - | 8 | 664 |