Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146077.2 | NP_666189.1 | 3 | 865 | ||
GenBank mRNA | AK033650.1 | - | 3 | 878 | ||
BC026666.1 | - | 3 | 865 | 4990292 |