Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146084.1 | NP_666196.1 | 6 | 1101 | ||
GenBank mRNA | AK029766.1 | - | 6 | 1093 | ||
BC031465.1 | - | 6 | 1101 | 3981146 |