Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146107.2 | NP_666219.1 | 11 | 1548 | ||
GenBank mRNA | AK010077.1 | - | 1 | 112 | ||
AK078508.1 | - | 11 | 1566 | |||
AK088839.1 | - | 11 | 1581 | |||
AK216909.1 | - | 1 | 230 | |||
BC023020.1 | - | 11 | 1548 | 5375425 | ||
BC024861.1 | - | 6 | 872 | |||
BC027134.1 | - | 2214 |