Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110212.2 | NP_001103682.1 | 5 | 442 | ||
NM_146112.4 | NP_666224.3 | 5 | 442 | |||
GenBank mRNA | AK079246.1 | - | 5 | 412 | ||
AK144058.1 | - | 5 | 404 | |||
AK160878.1 | - | 5 | 407 | |||
AY176043.1 | - | 5 | 388 | |||
BC021370.1 | - | 5 | 409 | |||
BC076610.1 | - | 5 | 388 | |||
BC089036.1 | - | 5 | 398 | 30695300 |