Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005792.2 | NP_005783.2 | 5 | 534 | ||
XM_011522808.2 | XP_011521110.1 | 6 | 814 | |||
XM_017022852.1 | XP_016878341.1 | 5 | 648 | |||
GenBank mRNA | AK222667.1 | - | 5 | 534 | ||
AW175781.1 | - | 5 | 566 | |||
BC005242.1 | - | 5 | 505 | 3997566 | ||
BC011020.1 | - | 5 | 511 | 4287267 | ||
BC031017.1 | - | 5 | 506 | 4670801 | ||
BU595100.1 | - | 5 | 530 | |||
X98263.1 | - | 5 | 517 |