Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 5 | 691 | ||
GenBank mRNA | AK033735.1 | - | 5 | 659 | ||
AK046583.1 | - | 5 | 588 | |||
AK051396.1 | - | 5 | 630 | |||
AK146953.1 | - | 5 | 676 | |||
AK149818.1 | - | 5 | 602 | |||
AK149888.1 | - | 5 | 601 | |||
AK150642.1 | - | 5 | 629 | |||
AK152207.1 | - | 5 | 650 | |||
AK153489.1 | - | 5 | 635 | |||
AK170512.1 | - | 5 | 631 | |||
AK177466.1 | - | 2 | 136 | |||
AK206543.1 | - | 2 | 77 | |||
AK217216.1 | - | 2 | 77 | |||
BC027843.1 | - | 5 | 611 | 5343013 |