Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 8, 9 | 1186 | ||
GenBank mRNA | AK033735.1 | - | 8, 9 | 1154 | ||
AK046583.1 | - | 8, 9 | 1083 | |||
AK051396.1 | - | 8, 9 | 1125 | |||
AK146953.1 | - | 8, 9 | 1171 | |||
AK149818.1 | - | 8, 9 | 1097 | |||
AK149888.1 | - | 8, 9 | 1096 | |||
AK150642.1 | - | 8, 9 | 1124 | |||
AK152207.1 | - | 8, 9 | 1145 | |||
AK153489.1 | - | 8, 9 | 1130 | |||
AK170512.1 | - | 8, 9 | 1126 | |||
BC027843.1 | - | 8, 9 | 1106 | 5343013 |