Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 14 | 3282 | ||
GenBank mRNA | AK033735.1 | - | 14 | 3249 | ||
AK046583.1 | - | 14 | 3179 | |||
AK051396.1 | - | 14 | 3221 | |||
AK146953.1 | - | 14 | 3267 | |||
AK149818.1 | - | 14 | 3192 | |||
AK149888.1 | - | 14 | 3192 | |||
AK150642.1 | - | 14 | 3220 | |||
AK152207.1 | - | 14 | 3241 | |||
AK153489.1 | - | 14 | 3225 | |||
AK170512.1 | - | 14 | 3222 | |||
AK195145.1 | - | 1 | 100 | |||
BC024639.1 | - | 6 | 2013 | |||
BC027843.1 | - | 14 | 3200 | 5343013 |