Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146122.3 | NP_666234.3 | 4, 5 | 445 | ||
GenBank mRNA | AK137530.1 | - | 3, 4 | 373 | ||
BC027786.1 | - | 4, 5 | 408 | 5346095 | ||
BQ444119.1 | - | 4, 5 | 283 | |||
DQ448594.1 | - | 4, 5 | 190 |