Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146150.2 | NP_666262.2 | 1, 2 | 360 | ||
GenBank mRNA | AK031548.1 | - | 1, 2 | 360 | ||
AK166470.1 | - | 1, 2 | 487 | |||
BC026832.1 | - | 1, 2 | 410 | 4486176 | ||
BC036128.1 | - | 1, 2 | 478 | 4971791 |