Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039200.2 | NP_001034289.1 | 8 | 937 | ||
NM_146157.4 | NP_666269.2 | 8 | 937 | |||
GenBank mRNA | AK049061.1 | - | 8 | 928 | ||
AK077496.1 | - | 8 | 927 | |||
AK089289.1 | - | 8 | 916 | |||
AK138637.1 | - | 8 | 909 | |||
AK161677.1 | - | 8 | 900 | |||
AK170739.1 | - | 8 | 916 | |||
BC056952.1 | - | 3 | 385 | |||
BC139366.1 | - | 8 | 923 | 8862388 |