Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C87436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243741.1 | NP_001230670.1 | 10 | 2624 | ||
NM_001243742.1 | NP_001230671.1 | 10 | 2190 | |||
NM_146170.4 | NP_666282.3 | 10 | 2131 | |||
GenBank mRNA | AK047127.1 | - | 10 | 2132 | ||
AK077503.1 | - | 10 | 2511 | |||
AK170524.1 | - | 11 | 2022 | |||
AK171511.1 | - | 11 | 2022 | |||
AK189339.1 | - | 1 | 67 | |||
BC025632.1 | - | 10 | 2099 | 5321669 | ||
BC026926.1 | - | 11 | 2255 | 3586097 | ||
BC031913.1 | - | 10 | 2144 | 5376246 |