Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301407.1 | NP_001288336.1 | 8 | 1283 | ||
NM_146173.3 | NP_666285.1 | 8 | 1286 | |||
GenBank mRNA | AF276890.1 | - | 8 | 1285 | ||
AK004954.1 | - | 8 | 1259 | |||
AK172176.1 | - | 8 | 1264 | |||
BC024685.1 | - | 8 | 1244 | 3981492 |