Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ffar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168509.1 | NP_001161981.1 | 3 | 1454 | ||
NM_001168510.1 | NP_001161982.1 | 3 | 1391 | |||
NM_001168511.1 | NP_001161983.1 | 3 | 1398 | |||
NM_001168512.1 | NP_001161984.1 | 2 | 1544 | |||
NM_146187.4 | NP_666299.1 | 3 | 1777 | |||
GenBank mRNA | AK078861.1 | - | 3 | 1781 |